Learn More
Biomatik Corporation Human Zinc transporter 8 (SLC30A8) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36081
Encompass_Preferred
Human Zinc Transporter 8 (Slc30A8) Elisa Kit for detecting Solute Carrier Family 30 (Zinc Transporter), Member 8 in serum, plasma, cell lysates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 80-1400pg/ml. Sensitivity: 50pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8IWU4; SLC30A8. Target Synonyms: SLC30A8; ZNT8; Zinc transporter 8; ZnT-8; Solute carrier family 30 member 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8361
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.