Learn More
Biomatik Corporation Monkey Vascular Endothelial cell Growth Factor (VEGF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36204
Encompass_Preferred
Monkey Vascular Endothelial Cell Growth Factor (Vegf) Elisa Kit for detecting Vascular Endothelial Cell Growth Factor(Vegf) in serum, plasma, cell culture supernates, cell lysates. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8392