Learn More
Biomatik Corporation Mouse Heat shock protein HSP 90-beta (HSP90AB1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37028
Encompass_Preferred
Mouse Heat Shock Protein Hsp 90-Beta (Hsp90Ab1) Elisa Kit for detecting Heat Shock Protein 90Kda Alpha (Cytosolic), Class B Member 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P11499; Hsp90ab1. Target Synonyms: Hsp90ab1; Hsp84; Hsp84-1; Hspcb; Heat shock protein HSP 90-beta; Heat shock 84 kDa; HSP 84; HSP84; Tumor-specific transplantation 84 kDa antigen; TSTA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8531