Learn More
Biomatik Corporation Pig calcitonin gene related peptide (CGRP) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKC38114
Pig Calcitonin Gene Related Peptide (Cgrp) Elisa Kit for detecting Calcitonin Gene Related Peptide(Cgrp) in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: A6P7L6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.