Learn More
Biomatik Corporation Pig Collagen Type , Col ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38143
Encompass_Preferred
Pig Collagen Type Iii, Col Iii Elisa Kit for detecting Collagen Type III, Col III in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 6.25-400ng/ml. Sensitivity: 1.56ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: Collagen Type 3; Col 3; Collagen Type III; Col III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8726
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.