Learn More
Biomatik Corporation Pig glial fibrillary acidic protein, GFAP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38192
Encompass_Preferred
Pig Glial Fibrillary Acidic Protein, Gfap Elisa Kit for detecting Glial Fibrillary Acidic Protein, Gfap in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.1-20ng/ml. Sensitivity: 0.05ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: F1RR02; GFAP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8737
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.