Learn More
Biomatik Corporation Pig insulin-like growth factors-1, IGF-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38227
Encompass_Preferred
Pig Insulin-Like Growth Factors-1, Igf-1 Elisa Kit for detecting Insulin-Like Growth Factor 1 (Somatomedin C) in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 6.25-400ng/ml. Sensitivity: 1.56ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P16545; IGF1. Target Synonyms: IGF1; Insulin-like growth factor I; IGF-I; Somatomedin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8744
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.