Learn More
Biomatik Corporation Rabbit Collagen Type , Col ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38415
Encompass_Preferred
Rabbit Collagen Type I, Col I Elisa Kit for detecting Collagen Type I, Col I in serum, plasma, tissue homogenates. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: Collagen Type 1; Col 1; Collagen Type I; Col I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8780
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.