Learn More
Biomatik Corporation Rat Glutathione Peroxidase (GSH-PX) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39116
Encompass_Preferred
Rat Glutathione Peroxidase (Gsh-Px) Elisa Kit for detecting Glutathione Peroxidase 1 in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.5-800mIU/ml. Sensitivity: 3.12mIU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04041; Gpx1. Target Synonyms: Gpx1Glutathione peroxidase 1; GPx-1; GSHPx-1; EC 1.11.1.9; Cellular glutathione peroxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8883
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.