Learn More
Biomatik Corporation Rat Nuclear factor-kappa B (NF-B) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39499
Encompass_Preferred
Rat Nuclear Factor-Kappa B (Nf-Κb) Elisa Kit for detecting Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells 1 in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q63369; Nfkb1. Target Synonyms: Nfkb1; Nuclear factor NF-kappa-B p105 subunit; DNA-binding factor KBF1; EBP-1; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1) [Cleaved into: Nuclear factor NF-kappa-B p50 subunit]; Fragment. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8949
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.