Learn More
Biomatik Corporation Rat tartrate-resistant acid phosphatase 5b, TRACP-5b ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39744
Encompass_Preferred
Rat Tartrate-Resistant Acid Phosphatase 5B, Tracp-5B Elisa Kit for detecting Tartrate-Resistant Acid Phosphatase 5B, Tracp-5B in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20mIU/ml. Sensitivity: 0.078mIU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8993
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.