Learn More
Biomatik Corporation Rat Angiotensin converting enzyme, ACE ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40025
Encompass_Preferred
Rat Angiotensin Converting Enzyme, Ace Elisa Kit for detecting Angiotensin I Converting Enzyme (Peptidyl-Dipeptidase A) 1 in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000ng/ml. Sensitivity: 7.81ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P47820; Ace. Target Synonyms: Ace; Dcp1; Angiotensin-converting enzyme; ACE; EC 3.2.1.-; EC 3.4.15.1; Dipeptidyl carboxypeptidase I; Kininase II; CD antigen CD143) [Cleaved into: Angiotensin-converting enzyme; soluble form]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9026
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.