Learn More
Biomatik Corporation Human brain natriuretic peptide, BNP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40359
Encompass_Preferred
The Human brain natriuretic peptide, BNP ELISA Kit allows for quantitative determination of brain natriuretic peptide, BNP in serum, urine, tissue homogenates, cell culture supernates. Species Reactivity: Human. Detection Range: 93.75 pg/mL-6000 pg/mL. Sensitivity: 23.43 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-9164
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.