Learn More
Biomatik Corporation Human B cell activation factor,BAFF ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40831
Encompass_Preferred
The Human B cell activation factor,BAFF ELISA Kit allows for quantitative determination of tumor necrosis factor (ligand) superfamily, member 13b in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9Y275. Restriction: For Research Use Only.
Catalog No. 50-149-9339
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.