Learn More
Biomatik Corporation Rat Keratinocyte Growth Factor, KGF ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40900
Encompass_Preferred
Rat Keratinocyte Growth Factor, Kgf Elisa Kit for detecting Fibroblast Growth Factor 7 (Keratinocyte Growth Factor) in serum, plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q02195; Fgf7. Target Synonyms: Fgf7; Fgf-7; KgfFibroblast growth factor 7; FGF-7; Heparin-binding growth factor 7; HBGF-7; Keratinocyte growth factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9368
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.