Learn More
Biomatik Corporation Rat KI-67 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40955
Encompass_Preferred
Rat Ki-67 Elisa Kit for detecting Ki-67 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.04ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q5RJM0; Nifk. Target Synonyms: Nifk; Mki67ipMKI67 FHA domain-interacting nucleolar phosphoprotein; Nucleolar protein interacting with the FHA domain of pKI-67. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9392
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.