Learn More
Biomatik Corporation Monkey estradiol (E2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41220
Encompass_Preferred
Monkey Estradiol (E2) Elisa Kit for detecting Estradiol (E2) in serum, plasma. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 40-1000pg/ml. Sensitivity: 40pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9477
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.