Learn More
Biomatik Corporation Human Corin ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41293
Encompass_Preferred
The Human Corin ELISA Kit allows for quantitative determination of corin, serine peptidase in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human. Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9Y5Q5. Restriction: For Research Use Only.
Catalog No. 50-149-9495
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.