Learn More
Biomatik Corporation Mouse Cadherin-5 (CDH5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41435
Encompass_Preferred
Mouse Cadherin-5 (Cdh5) Elisa Kit for detecting Cadherin 5, Type 2 (Vascular Endothelium) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.235-15ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P55284; Cdh5. Target Synonyms: Cdh5; Cadherin-5; Vascular endothelial cadherin; VE-cadherin; CD antigen CD144. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9526
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.