Learn More
Biomatik Corporation Human Calcium-dependent phospholipase A2 (PLA2G5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41838
Encompass_Preferred
The Human Calcium-dependent phospholipase A2 (PLA2G5) ELISA Kit allows for quantitative determination of phospholipase A2, group V in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 0.18 ng/mL-12 ng/mL. Sensitivity: 0.04 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P39877. Restriction: For Research Use Only.
Catalog No. 50-149-9599
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.