Learn More
Biomatik Corporation Human SLAM family member 7 (SLAMF7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41928
Encompass_Preferred
Human Slam Family Member 7 (Slamf7) Elisa Kit for detecting Slam Family Member 7 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NQ25; SLAMF7. Target Synonyms: SLAMF7; CS1; UNQ576/PRO1138; SLAM family member 7; CD2 subset 1; CD2-like receptor-activating cytotoxic cells; CRACC; Membrane protein FOAP-12; Novel Ly9; Protein 19A; CD antigen CD319. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9618
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.