Learn More
Biomatik Corporation Human BK (Bradykinin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60408
Encompass_Preferred
The Human BK (Bradykinin) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 1.56-100 ng/mL. Sensitivity: 0.94 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive. Detection Method: Colorimetric. Assay Time: 2.5 hours. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-149-9941
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.