Learn More
Biomatik Corporation Human SDC1 (Syndecan 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60804
Encompass_Preferred
Human SDC1 (Syndecan 1) ELISA Kit for detecting SDC1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.16-10ng/ml. Sensitivity: 0.1ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P18827. Target Synonyms: CD138; SDC; SYND1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0190
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.