Learn More
Biomatik Corporation Human Trx (Thioredoxin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE61034
Encompass_Preferred
Human Trx (Thioredoxin) ELISA Kit for detecting Trx in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.31-20ng/ml. Sensitivity: 0.19ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P10599. Target Synonyms: TXN; TRDX; TRX1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0367
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.