Learn More
Biomatik Corporation Human IL-27 (Interleukin 27) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKE61275
The Human IL-27 (Interleukin 27) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 31.25-2000 pg/mL. Sensitivity: 18.75 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: Q8NEV9. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.