Learn More
Biomatik Corporation Human CK-MB (Creatine Kinase MB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57135
Encompass_Preferred
The Human CK-MB (Creatine Kinase MB) ELISA Kit allows for quantitative determination of CK-MB in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.094 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. Restriction: For Research Use Only.
Catalog No. 50-150-0781
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.