Learn More
Biomatik Corporation Rat IL-4 (Interleukin-4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57854
Encompass_Preferred
Rat IL-4 (Interleukin-4) ELISA Kit for detecting IL-4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P20096. Target Synonyms: IL-4; Interleukin 4; IL4; BCGF-1; BCGF1; BSF-1; BSF1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1026
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.