Learn More
Biomatik Corporation Estrone (Estrone) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57997
Encompass_Preferred
The Estrone (Estrone) ELISA Kit allows for quantitative determination of Estrone in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: All species. Detection Range: 0.156-10 pmol/mL. Sensitivity: 0.094 pmol/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive ELISA, Coated with Antigen. Restriction: For Research Use Only.
Catalog No. 50-150-1146
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.