Learn More
Biomatik Corporation Mouse Epidermal Growth Factor Receptor 2 (EGFR2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54232
Encompass_Preferred
Mouse Epidermal Growth Factor Receptor 2 (EGFR2) ELISA Kit for detecting EGFR2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.207ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P70424. Gene ID: 13866. Target Synonyms: CD340; ErbB2; HER-2/neu; HER2; NEU; MLN 19; NGL; TKR1; p185; V-Erb-B2 Erythroblastic Leukemia Viral Oncogene Homolog 2; Neuro/Glioblastoma Derived Oncogene Homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1233
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.