Learn More
Biomatik Corporation Human Pleiotrophin (PTN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54898
Encompass_Preferred
The Human Pleiotrophin (PTN) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 7.1 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P21246. NCBI Gene ID: 5764. Restriction: For Research Use Only.
Catalog No. 50-150-1557
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.