Learn More
Biomatik Corporation Human Transforming Growth Factor Beta 3 (TGFb3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54980
Encompass_Preferred
Human Transforming Growth Factor Beta 3 (TGFb3) ELISA Kit for detecting TGFb3 in serum, plasma, tissue homogenates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 34pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P10600. Gene ID: 7043. Target Synonyms: TGF-B3; LAP; Latency-associated peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1586
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.