Learn More
Biomatik Corporation Human Adropin (AD) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02192
Encompass_Preferred
The Human Adropin (AD) ELISA Kit allows for quantitative determination of Adropin in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 13.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q6UWT2. Cross Activity: No significant cross-reactivity or interference between Adropin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1718
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.