Learn More
Biomatik Corporation Androsterone (ADT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02347
Encompass_Preferred
The Androsterone (ADT) ELISA Kit allows for quantitative determination of Androsterone in Serum, Plasma and other biological fluids. Species Reactivity: All species. Detection Range: 352-90000 pg/mL. Sensitivity: 144 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Androsterone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1785
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.