Learn More
Biomatik Corporation Dog Brain Natriuretic Peptide (BNP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02793
Encompass_Preferred
The Dog Brain Natriuretic Peptide (BNP) ELISA Kit allows for quantitative determination of Brain Natriuretic Peptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 24.69-2,000 pg/mL. Sensitivity: 9.55 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Brain Natriuretic Peptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1957
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.