Learn More
Biomatik Corporation Rabbit C Reactive Protein (CRP) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU02827
Rabbit C Reactive Protein (CRP) ELISA Kit for detecting CRP in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.28ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02742. Gene ID: 100009091. Target Synonyms: C-RP; PTX1; Pentraxin-Related; Pentraxin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.