Learn More
Biomatik Corporation Human Collagen Type II Alpha 1 (COL2a1) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU03312
The Human Collagen Type II Alpha 1 (COL2a1) ELISA Kit allows for quantitative determination of Collagen Type II Alpha 1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 12.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02458. Cross Activity: No significant cross-reactivity or interference between Collagen Type II Alpha 1 and analogues was observed. Restriction: For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.