Learn More
Biomatik Corporation Mouse Connective Tissue Growth Factor (CTGF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03437
Encompass_Preferred
Mouse Connective Tissue Growth Factor (CTGF) ELISA Kit for detecting CTGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29268. Gene ID: 14219. Target Synonyms: CCN2; HCS24; IGFBP8; NOV2; Insulin-Like Growth Factor-Binding Protein 8; Hypertrophic Chondrocyte-Specific Protein 24; CCN Family Member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2232
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.