Learn More
Biomatik Corporation Human Desmoglein 2 (DSG2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03709
Encompass_Preferred
The Human Desmoglein 2 (DSG2) ELISA Kit allows for quantitative determination of Desmoglein 2 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.054 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q14126. Cross Activity: No significant cross-reactivity or interference between Desmoglein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2337
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.