Learn More
Biomatik Corporation Dihydrotestosterone (DHT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03743
Encompass_Preferred
The Dihydrotestosterone (DHT) ELISA Kit allows for quantitative determination of Dihydrotestosterone in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: All species. Detection Range: 30.9-2500 pg/mL. Sensitivity: 13.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Dihydrotestosterone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2353
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.