Learn More
Biomatik Corporation Human Fatty Acid Synthase (FASN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04053
Encompass_Preferred
The Human Fatty Acid Synthase (FASN) ELISA Kit allows for quantitative determination of Fatty Acid Synthase in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.131 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P49327. Cross Activity: No significant cross-reactivity or interference between Fatty Acid Synthase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2456