Learn More
Biomatik Corporation Gamma-Aminobutyric Acid (gABA) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU04330
The Gamma-Aminobutyric Acid (gABA) ELISA Kit allows for quantitative determination of Gamma-Aminobutyric Acid in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: All species. Detection Range: 24.69-2,000 pg/mL. Sensitivity: 9.27 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Gamma-Aminobutyric Acid and analogues was observed. Restriction: For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.