Learn More
Biomatik Corporation Human C-Met (MET) ELISA Kit
Supplier: Biomatik Corporation EKU04756
Human C-Met (MET) ELISA Kit for detecting MET in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8,000pg/ml. Sensitivity: 49pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08581. Gene ID: 4233. Target Synonyms: HGFR; RCCP2; Hepatocyte Growth Factor Receptor; Met Proto-Oncogene; Mesenchymal-Epithelial Transition Factor; HGF/SF receptor; Proto-oncogene c-Met; Scatter factor receptor; Tyrosine-protein kinase M. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.