Learn More
Biomatik Corporation Gibberellic Acid (GA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04370
Encompass_Preferred
The Gibberellic Acid (GA) ELISA Kit allows for quantitative determination of Gibberellic Acid in tissue or Cell culture supernates. Species Reactivity: All species. Detection Range: 123.5-10,000 ng/mL. Sensitivity: 52.1 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Gibberellic Acid and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2574
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.