Learn More
Biomatik Corporation Mouse Orexin (OX) ELISA Kit
Supplier:  Biomatik Corporation EKU06397
Encompass_Preferred
Mouse Orexin (OX) ELISA Kit for detecting OX in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.17-500pg/ml. Sensitivity: 2.79pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: O55241. Gene ID: 15171. Target Synonyms: HCRT; PPOX; Orexin A; Hypocretin 1; Orexin B; Hypocretin 2; Preproorexin; Hypocretin Neuropeptide Precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6284