Learn More
Biomatik Corporation Mouse Granzyme K (GZMK) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04569
Encompass_Preferred
Mouse Granzyme K (GZMK) ELISA Kit for detecting GZMK in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35205. Gene ID: 14945. Target Synonyms: GZM-K; TRYP2; PRSS; Serine Protease,Granzyme 3; Tryptase II; Fragmentin-3; Granzyme-3; NK-tryptase-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2651
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.