Learn More
Biomatik Corporation Prostaglandin F2 Alpha (PGF2a) ELISA Kit
Supplier: Biomatik Corporation EKU06864
Prostaglandin F2 Alpha (PGF2a) ELISA Kit for detecting PGF2a in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 9.88-800pg/ml. Sensitivity: 4.13pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: PGF2A; PGF2-A; PG-F2a. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.