Learn More
Biomatik Corporation Fish Prolactin/Luteotropic Hormone (Prl/Lth) Elisa Kit
Supplier:  Biomatik Corporation EKC32166
Encompass_Preferred
Fish Prolactin/Luteotropic Hormone (Prl/Lth) Elisa Kit for detecting Prolactin/Luteotropic Hormone(Prl/Lth) in serum, plasma, tissue homogenates. Species Reactivity: Fish (Vertebrata). Detection Range: 10-4000uIU/ml. Sensitivity: 10uIU/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P09319; prl1. Target Synonyms: prl1; Prolactin-1; PRL-188; Prolactin I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7275
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.