Learn More
Biomatik Corporation Hamster Interleukin 6, Il-6 Elisa Kit
Supplier:  Biomatik Corporation EKC32293
Encompass_Preferred
Hamster Interleukin 6, Il-6 Elisa Kit for detecting Interleukin 6, Il-6 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Golden hamster. Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7291
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.