Learn More
Biomatik Corporation Gallus Interferon Gamma (IFNg) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05099
Encompass_Preferred
The Gallus Interferon Gamma (IFNg) ELISA Kit allows for quantitative determination interferon Gamma in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P49708. Cross Activity: No significant cross-reactivity or interference between Interferon Gamma and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2875
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.