Learn More
Biomatik Corporation Mouse Interleukin 12A (IL12A) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU05202
Mouse Interleukin 12A (IL12A) ELISA Kit for detecting IL12A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43431. Gene ID: 16159. Target Synonyms: p35; IL12-A; CLMF1; NFSK; NKSF1; Natural Killer Cell Stimulatory Factor 1; Cytotoxic Lymphocyte Maturation Factor 1; NF Cell Stimulatory Factor Chain 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.